3-Amino-4-methoxybenzamide ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151420-100G A151420-25G A151420-5G
ALA151420-100GEA 185.16 USD
ALA151420-100G ALA151420-25G ALA151420-5G
3-Amino-4-methoxybenzamide ≥98%
3-Amino-4-methoxybenzamide
Formula: C₈H₁₀N₂O₂
Melting Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00017132
CAS Number: 17481-27-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0598303429f4f5c6f48bc5399c78fc8ab0ec435&vid=b0598303429f4f5c6f48bc5399c78fc8ab0ec435&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR