5-Acetylsalicylamide 98

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 5-Acetyl-2-hydroxybenzamide

A151418-25G A151418-5G
ALA151418-25GEA 40.76 USD
ALA151418-25G ALA151418-5G
5-Acetylsalicylamide 98
5-Acetylsalicylamide
Formula: C₉H₉NO₃
MW: 179.18 g/mol
Melting Pt: 224 °C
MDL Number: MFCD00049222
CAS Number: 40187-51-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07db2807b1525fa6695b63847858eacc7c0c2cce&vid=07db2807b1525fa6695b63847858eacc7c0c2cce&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR