4-(4-Chlorophenoxy)aniline ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A151417-25G
A151417-50G
A151417-5G
ALA151417-25GEA
142.41
USD
ALA151417-25G
ALA151417-50G
ALA151417-5G
4-(4-Chlorophenoxy)aniline ≥98% (by GC)
4-(4-Chlorophenoxy)aniline
Formula:
C₁₂H₁₀ClNO Boiling Pt: 205 °C Melting Pt: 101 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00043925 CAS Number: 101-79-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...