4-Amidinobenzamide hydrochloride ≥92% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Amidinobenzamide Hydrochloride, 4-Carbamimidoylbenzamide Hydrochloride

A151415-1G A151415-5G
ALA151415-1GEA 114.86 USD
ALA151415-1G ALA151415-5G
4-Amidinobenzamide hydrochloride ≥92% (by HPLC)
4-Amidinobenzamide hydrochloride
Formula: C₈H₉N₃O
Storage Temperature: Refrigerator
MDL Number: MFCD00013011
CAS Number: 59855-11-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8864ce690461403d2d6d16c20bb112e11a758b7&vid=e8864ce690461403d2d6d16c20bb112e11a758b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR