Adipamide ≥98% (by total nitrogen basis)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A151413-1G A151413-25G A151413-5G
ALA151413-1GEA 9.41 USD
ALA151413-1G ALA151413-25G ALA151413-5G
Adipamide ≥98% (by total nitrogen basis)
Adipamide

Adipamide has been used in the preparation of bisimidates

Formula: C₆H₁₂N₂O₂
MW: 144.17 g/mol
Melting Pt: 224-228°C
Storage Temperature: Ambient
MDL Number: MFCD00008045
CAS Number: 628-94-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6d61efd2da5e2591ce55df35d1b312853f30176&vid=c6d61efd2da5e2591ce55df35d1b312853f30176&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR