6-Azathymine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 6-Methyl-1,2,4-triazine-3,5(2H,4H)-dione

A151412-1G A151412-250MG A151412-25G A151412-5G
ALA151412-1GEA 403.66 USD
ALA151412-1G ALA151412250MG ALA151412-25G ALA151412-5G
6-Azathymine ≥97%
6-Azathymine
Formula: C₄H₅N₃O₂
MW: 127.10 g/mol
Melting Pt: 214 °C
MDL Number: MFCD00006457
CAS Number: 932-53-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a172ac1f1f88c29c29c1cb4770bcec052c686ea2&vid=a172ac1f1f88c29c29c1cb4770bcec052c686ea2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR