N-Acetylsulphanilamide ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: N4-Acetylsulphanilamide, 4-Acetamidobenzenesulphonamide

A151411-1G A151411-25G A151411-5G
ALA151411-1GEA 9.41 USD
ALA151411-1G ALA151411-25G ALA151411-5G
N-Acetylsulphanilamide ≥98% (by HPLC)
N-Acetylsulphanilamide
Formula: C₈H₁₀N₂O₃S
MW: 214.2416 g/mol
Melting Pt: 216 °C
MDL Number: MFCD00035784
CAS Number: 121-61-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc6f5ad301c72c84b5378e11ea8848f9362a90b4&vid=dc6f5ad301c72c84b5378e11ea8848f9362a90b4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR