2-(Aminomethyl)-5-methylpyrazine ≥97% (by GC, titration analysis)

Supplier: Aladdin Scientific
Danger

A151407-5G A151407-1G A151407-250MG
ALA151407-5GEA 297.26 USD
ALA151407-5G ALA151407-1G ALA151407250MG
2-(Aminomethyl)-5-methylpyrazine ≥97% (by GC, titration analysis)
2-(Aminomethyl)-5-methylpyrazine
Formula: C₆H₉N₃
Boiling Pt: 94 °C
Density: 1.09 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD01314542
CAS Number: 132664-85-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9e93f27dd08ad5f23d05a56c511c1ca0510218e&vid=f9e93f27dd08ad5f23d05a56c511c1ca0510218e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR