4-Aminoazobenzene hydrochloride ≥98% (by titration)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 4-(Phenylazo)aniline Hydrochloride, 4-Aminoazobenzene Hydrochloride

A151403-100G A151403-25G
ALA151403-100GEA 198.46 USD
ALA151403-100G ALA151403-25G
4-Aminoazobenzene hydrochloride ≥98% (by titration)
4-Aminoazobenzene hydrochloride
Formula: C₁₂H₁₁N₃
Melting Pt: 213 °C
MDL Number: MFCD00012994
CAS Number: 3457-98-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a78b77a20caa0f1e99db878ad96a30f0b7858f65&vid=a78b77a20caa0f1e99db878ad96a30f0b7858f65&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR