4-Acetylbenzoic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151401-100G A151401-25G A151401-1G
ALA151401-100GEA 1579.76 USD
ALA151401-100G ALA151401-25G ALA151401-1G
4-Acetylbenzoic acid ≥98%
4-Acetylbenzoic acid
Formula: C₉H₈O₃
Melting Pt: 210 °C
MDL Number: MFCD00002561
CAS Number: 586-89-0
EINECS: 209-588-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d65322979f07740573859fa1c3ad264f770d6278&vid=d65322979f07740573859fa1c3ad264f770d6278&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR