1-Allylimidazole 97 GC_ASSAY_METHOD

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 1-Allyl-1H-imidazole

A151399-100G A151399-25G A151399-5G
ALA151399-100GEA 146.21 USD
ALA151399-100G ALA151399-25G ALA151399-5G
1-Allylimidazole 97 GC_ASSAY_METHOD
1-Allylimidazole
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 212-213℃
Flash Pt: 101°C
MDL Number: MFCD00037894
CAS Number: 31410-01-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3409f3e7dd382f757258019647ad097d37fbaa1f&vid=3409f3e7dd382f757258019647ad097d37fbaa1f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR