3-Acetyl-2,5-dimethylfuran ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 1-(2,5-Dimethylfuran-3-yl)ethanone

A151396-25G A151396-5G
ALA151396-25GEA 75.91 USD
ALA151396-25G ALA151396-5G
3-Acetyl-2,5-dimethylfuran ≥98% (by GC)
3-Acetyl-2,5-dimethylfuran

Product Introduction:3-Acetyl-2,5-dimethylfuran is a flavouring agent and its specifications were revised.Application3-Acetyl-2,5-dimethylfuran was used in synthesis of:furylfulgide, 2-1-(2, 5-dimethyl-3-furyl) ethylidene-3-isopropylidene succinic anhydrideChalcones1-(2′,5′-dimethyl-3′-furyl)-3-(aryl)-2-propen-1-one

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 196°C
Density: 1.04 g/cm³
Flash Pt: 82°C
MDL Number: MFCD00010281
CAS Number: 10599-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11e30f82a5f330141e77fd468d6bab3d288b70b0&vid=11e30f82a5f330141e77fd468d6bab3d288b70b0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR