1-Acetyl-5-bromoindoline ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
1-(5-Bromoindolin-1-yl)ethanone
A151395-1G
A151395-25G
A151395-5G
ALA151395-1GEA
28.41
USD
ALA151395-1G
ALA151395-25G
ALA151395-5G
1-Acetyl-5-bromoindoline ≥98% (by GC)
1-Acetyl-5-bromoindoline
Formula:
C₁₀H₁₀BrNO Melting Pt: 121 °C Storage Temperature: Freezer |
MDL Number:
MFCD00056017 CAS Number: 22190-38-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...