2-Aminoazotoluene hydrochloride ≥98% (by titration)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4'-Amino-2,3'-dimethylazobenzene Hydrochloride, 2-Aminoazotoluene Hydrochloride

A151392-100G A151392-25G A151392-5G
ALA151392-100GEA 327.66 USD
ALA151392-100G ALA151392-25G ALA151392-5G
2-Aminoazotoluene hydrochloride ≥98% (by titration)
2-Aminoazotoluene hydrochloride
Formula: C₁₄H₁₆ClN₃
Melting Pt: >300°C
MDL Number: MFCD00060210
CAS Number: 2298-13-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e63d901e203272d7cf39c3c8c8d529e76d383dc&vid=8e63d901e203272d7cf39c3c8c8d529e76d383dc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR