2-Aminoethyl hydrogen sulfate ≥98% (by titration)

Supplier: Aladdin Scientific
Warning

A151387-100G A151387-25G A151387-5G
ALA151387-100GEA 84.46 USD
ALA151387-100G ALA151387-25G ALA151387-5G
2-Aminoethyl hydrogen sulfate ≥98% (by titration)
2-Aminoethyl hydrogen sulphate
Formula: C₂H₇NO₄S
Melting Pt: 279 °C
MDL Number: MFCD00008179
CAS Number: 926-39-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6ec466324c0857c7f55f8887f891923171e9fb1&vid=f6ec466324c0857c7f55f8887f891923171e9fb1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR