1-(Azidomethyl)pyrene ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC

A151381-100MG A151381-20MG A151381-500MG
ALA151381100MGEA 94.91 USD
ALA151381100MG ALA151381-20MG ALA151381500MG
1-(Azidomethyl)pyrene ≥98% (by HPLC)
1-(Azidomethyl)pyrene
Formula: C₁₇H₁₁N₃
Melting Pt: 68 °C
Storage Temperature: Freezer
CAS Number: 1006061-57-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=846d68ab91aa29dd9e5ae8516a8a573e0765e892&vid=846d68ab91aa29dd9e5ae8516a8a573e0765e892&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR