1-Anthracenecarboxylic acid ≥99%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-Anthracenecarboxylic Acid

A151377-250MG A151377-100MG A151377-1G
ALA151377250MGEA 95.86 USD
ALA151377250MG ALA151377100MG ALA151377-1G
1-Anthracenecarboxylic acid ≥99%
1-Anthracenecarboxylic acid
Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
Melting Pt: 252 °C
MDL Number: MFCD00142583
CAS Number: 607-42-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59d70599ca500ba7e7f205dac6c82d3eee4c6da5&vid=59d70599ca500ba7e7f205dac6c82d3eee4c6da5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR