9,10-Anthraquinone-2-carboxylic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Anthraquinone-2-carboxylic acid, 9,10-Dioxo-9,10-dihydroanthracene-2-carboxylic acid

A151376-100G A151376-1G A151376-25G A151376-5G
ALA151376-100GEA 1036.36 USD
ALA151376-100G ALA151376-1G ALA151376-25G ALA151376-5G
9,10-Anthraquinone-2-carboxylic acid ≥98%
9,10-Anthraquinone-2-carboxylic acid

Application:Anthraquinone 2-carboxylic acid (AQ) as a novel electron shuttling mediator and attached electron relay for HRP. Efficient method for the generation of hydrogen peroxide by aerobic photooxidation of 2-propanol using anthraquinone-2-carboxylic acid and molecular oxygen is investigated.

Formula: C₁₅H₈O₄
MW: 252.22 g/mol
Melting Pt: 287-289°C
MDL Number: MFCD00001231
CAS Number: 117-78-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37f8a0c2949eb9bd7e5043d89ae0fd325bac543e&vid=37f8a0c2949eb9bd7e5043d89ae0fd325bac543e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR