Pentylbenzene ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 1-Phenylpentane, Amylbenzene

A151374-100ML A151374-25ML A151374-5ML
ALA151374100MLEA 159.51 USD
ALA151374100ML ALA151374-25ML ALA151374-5ML
Pentylbenzene ≥98% (by GC)
Pentylbenzene
Formula: C₁₁H₁₆
MW: 148.25 g/mol
Boiling Pt: 205°C
Melting Pt: -75°C
Density: 0.86 g/cm³
Flash Pt: 66°C
MDL Number: MFCD00009502
CAS Number: 538-68-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=296d2142805c3338280c7c72392f49e622900c6f&vid=296d2142805c3338280c7c72392f49e622900c6f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR