ɑ-Cyclohexylphenylacetic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A151371-1G A151371-200MG A151371-25G A151371-5G
ALA151371-1GEA 36.96 USD
ALA151371-1G ALA151371200MG ALA151371-25G ALA151371-5G
ɑ-Cyclohexylphenylacetic acid ≥95%
alpha-Cyclohexylphenylacetic acid
Formula: C₁₄H₁₈O₂
Melting Pt: 152 °C
Storage Temperature: Freezer
MDL Number: MFCD00001477
CAS Number: 3894-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR