5-Chloro-2-(4-chlorophenoxy)aniline ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-4,4'-dichlorodiphenyl ether, 5-Chloro-2-(4-chlorophenoxy)aniline

A151367-1G A151367-250MG A151367-25G A151367-5G
ALA151367-1GEA 24.61 USD
ALA151367-1G ALA151367250MG ALA151367-25G ALA151367-5G
5-Chloro-2-(4-chlorophenoxy)aniline ≥98%
5-Chloro-2-(4-chlorophenoxy)aniline
Formula: C₁₂H₉Cl₂NO
MW: 254.1100 g/mol
Boiling Pt: 234°C
Melting Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00027391
CAS Number: 121-27-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64f1961472bad7c27baf74c9743ec7d87e91f75f&vid=64f1961472bad7c27baf74c9743ec7d87e91f75f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR