2-(4-Chlorophenoxy)aniline ≥97%
Supplier: ALADDIN SCIENTIFIC
A151364-1G
A151364-250MG
ALA151364-1GEA
50.26
USD
ALA151364-1G
ALA151364250MG
2-(4-Chlorophenoxy)aniline ≥97%
2-(4-Chlorophenoxy)aniline
Formula:
C₁₂H₁₀ClNO Boiling Pt: 202°C Density: 1.246 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00025168 CAS Number: 2770-11-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...