5-Amino-2,3-dicyano-1,4-naphthoquinone ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A151363-100MG A151363-250MG
ALA151363100MGEA 127.21 USD
ALA151363100MG ALA151363250MG
5-Amino-2,3-dicyano-1,4-naphthoquinone ≥97%
5-Amino-2,3-dicyano-1,4-naphthoquinone
Formula: C₁₂H₅N₃O₂
MW: 223.19 g/mol
MDL Number: MFCD00671485
CAS Number: 68217-29-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b41ab77cfce0bd5d2d2fc1a668b2138f7f6ee799&vid=b41ab77cfce0bd5d2d2fc1a668b2138f7f6ee799&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR