Amyl methyl sulfide ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A151354-1ML A151354-25ML A151354-5ML
ALA151354-1MLEA 31.26 USD
ALA151354-1ML ALA151354-25ML ALA151354-5ML
Amyl methyl sulfide ≥97%
Amyl methyl sulfide
Formula: C₆H₁₄S
Boiling Pt: 145°C
Melting Pt: -94°C
Density: 0.84 g/cm³
Flash Pt: 40 °C
MDL Number: MFCD00027274
CAS Number: 1741-83-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c02f80872115923149116fa702d068da2a97aaaa&vid=c02f80872115923149116fa702d068da2a97aaaa&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR