2-Aminopentane ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-Amylamine, 1-Methylbutylamine, 2-Pentylamine

A151353-1G A151353-25G A151353-5G
ALA151353-1GEA 9.41 USD
ALA151353-1G ALA151353-25G ALA151353-5G
2-Aminopentane ≥97% (by GC)
2-Aminopentane
Formula: C₅H₁₃N
MW: 87.17 g/mol
Boiling Pt: 91°C
Melting Pt: 90.5-91.5°C
Density: 0.736 g/cm³
Flash Pt: -6°C
Storage Temperature: Ambient
MDL Number: MFCD00008098
CAS Number: 63493-28-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2699c0387ea4939c082feacbc8a24b4f0c9dfd90&vid=2699c0387ea4939c082feacbc8a24b4f0c9dfd90&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR