Anthracene-2,6-diyl bis(trifluoromethanesulfonate) ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
2,6-Bis(trifluoromethanesulphonyloxy)anthracene, Anthracene-2,6-diyl Bis(trifluoromethanesulphonate), 2,6-Bis(trifluoromethanesulfonyloxy)anthracene
A151345-250MG
A151345-50MG
ALA151345250MGEA
445.46
USD
ALA151345250MG
ALA151345-50MG
Anthracene-2,6-diyl bis(trifluoromethanesulfonate) ≥98%
Anthracene-2,6-diyl bis(trifluoromethanesulfonate)
Formula:
C₁₆H₈F₆O₆S₂ MW: 474.34 g/mol Melting Pt: 116-120°C Storage Temperature: Freezer |
CAS Number:
594838-61-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...