6-Acetoxy-7-methoxy-3H-quinazolin-4-one ≥98% (by HPLC, titration analysis)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 6-Acetoxy-7-methoxy-4-quinazolinol, 6-Acetoxy-7-methoxy-4(3H)-quinazolinone, 6-Acetoxy-7-methoxy-3,4-dihydroquinazolin-4-one, 6-Acetoxy-7-methoxy-4-hydroxyquinazoline

A151334-100G A151334-1G
ALA151334-100GEA 1408.76 USD
ALA151334-100G ALA151334-1G
6-Acetoxy-7-methoxy-3H-quinazolin-4-one ≥98% (by HPLC, titration analysis)
6-Acetoxy-7-methoxy-3H-quinazolin-4-one
Formula: C₁₁H₁₀N₂O₄
MW: 234.21 g/mol
Melting Pt: 297 °C
MDL Number: MFCD09951978
CAS Number: 179688-53-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4cd76888a9d9e17cf8c93fbb46b2796dd6fa535&vid=c4cd76888a9d9e17cf8c93fbb46b2796dd6fa535&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR