4-Amino-2-chloro-6,7-dimethoxyquinazoline ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
Doxazosin Related Compound C, 2-Chloro-4-amino-6,7-dimethoxyquinazoline
A151333-25G
A151333-5G
ALA151333-25GEA
207.96
USD
ALA151333-25G
ALA151333-5G
4-Amino-2-chloro-6,7-dimethoxyquinazoline ≥98% (by HPLC)
4-Amino-2-chloro-6,7-dimethoxyquinazoline
Formula:
C₁₀H₁₀ClN₃O₂ MW: 239.66 g/mol Melting Pt: 302 °C |
MDL Number:
MFCD00051734 CAS Number: 23680-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...