4,6-Diphenylpyrimidin-2-amine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-4,6-diphenylpyrimidine

A151332-1G A151332-250MG A151332-5G
ALA151332-1GEA 98.71 USD
ALA151332-1G ALA151332250MG ALA151332-5G
4,6-Diphenylpyrimidin-2-amine ≥98%
4,6-Diphenylpyrimidin-2-amine
Formula: C₁₆H₁₃N₃
MW: 247.30 g/mol
Melting Pt: 136 °C
MDL Number: MFCD00234910
CAS Number: 40230-24-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b033b45ae4e064a7af52b30ce863cc7b6433556&vid=4b033b45ae4e064a7af52b30ce863cc7b6433556&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR