(4-Amino-2-methyl-5-pyrimidinyl)methanol ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Toxopyrimidine, 4-Amino-2-methyl-5-pyrimidinemethanol, 4-Amino-5-hydroxymethyl-2-methylpyrimidine

A151329-1G A151329-250MG A151329-5G A151329-100MG
ALA151329-1GEA 197.51 USD
ALA151329-1G ALA151329250MG ALA151329-5G ALA151329100MG
(4-Amino-2-methyl-5-pyrimidinyl)methanol ≥95%
(4-Amino-2-methyl-5-pyrimidinyl)methanol
Formula: C₆H₉N₃O
MW: 139.1552 g/mol
Melting Pt: 195.0 - 199.0 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00849239
CAS Number: 73-67-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc5123d9882f3dc930a01cb41f2d322d071c22fb&vid=fc5123d9882f3dc930a01cb41f2d322d071c22fb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR