4-Aminopyrimidine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A151327-1G A151327-250MG A151327-25G A151327-500MG A151327-5G
ALA151327-1GEA 18.91 USD
ALA151327-1G ALA151327250MG ALA151327-25G ALA151327500MG ALA151327-5G
4-Aminopyrimidine ≥98%
4-Aminopyrimidine

Product Introduction:The consequences of one-electron oxidation and one-electron reduction were studied for 4-aminopyrimidine.Application4-Aminopyrimidine has been used in the preparation of 1:4-dihydro-4-imino-1-methylpyrimidine hydriodide.

Formula: C₄H₅N₃
MW: 95.1 g/mol
Melting Pt: 147-152°C
Storage Temperature: Ambient
MDL Number: MFCD00006112
CAS Number: 591-54-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bde34b94737ccd7765eeb25ab6e6fad46d46bac&vid=7bde34b94737ccd7765eeb25ab6e6fad46d46bac&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR