4-Amino-2,6-dimethylpyrimidine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151326-1G A151326-25G A151326-5G A151326-100G
ALA151326-1GEA 22.71 USD
ALA151326-1G ALA151326-25G ALA151326-5G ALA151326-100G
4-Amino-2,6-dimethylpyrimidine ≥98% (by GC)
4-Amino-2,6-dimethylpyrimidine
Formula: C₆H₉N₃
Boiling Pt: 254°C
Melting Pt: 184-186°C
Storage Temperature: Ambient
MDL Number: MFCD00006106
CAS Number: 461-98-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c29a19451d7fda92657c8a1122c6bd9370af17f&vid=0c29a19451d7fda92657c8a1122c6bd9370af17f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR