α-Ionone ≥90% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151316-100G A151316-25G A151316-5G
ALA151316-100GEA 104.41 USD
ALA151316-100G ALA151316-25G ALA151316-5G
α-Ionone ≥90% (by GC)
α-Ionone

α-Ionone is one of the key volatile constituents of the oil from whitefly (Syngonium podophyllum Schott).

Formula: C₁₃H₂₀O
Boiling Pt: 148 °C
Density: 0.93 g/cm³
Flash Pt: 118 °C
MDL Number: MFCD00001565
CAS Number: 127-41-3
EINECS: 204-841-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85a4b4d1166471f3679057b23372f59fdae2137b&vid=85a4b4d1166471f3679057b23372f59fdae2137b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR