12-Aminolauric acid 98

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 12-Aminolauric Acid, 12-Aminododecylic acid, 12-Aminododecanoic acid, 12-Aminododecanoic acid

A151311-100G A151311-25G A151311-5G
ALA151311-100GEA 117.71 USD
ALA151311-100G ALA151311-25G ALA151311-5G
12-Aminolauric acid 98
12-Aminolauric acid

Application:peptide synthesis: suitable

Formula: C₁₂H₂₅NO₂
MW: 215.34 g/mol
Boiling Pt: 343
Melting Pt: 183
Flash Pt: 280 °C
MDL Number: MFCD00008153
CAS Number: 693-57-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8876c9c51df2b82da355f2fc306bc18cae6d549c&vid=8876c9c51df2b82da355f2fc306bc18cae6d549c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR