7-Aminoenanthic acid ≥98% (by titration)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 7-Aminoheptanoic acid, 7-Aminoheptylic acid

A151309-100G A151309-1G A151309-250MG A151309-25G A151309-5G
ALA151309-100GEA 546.16 USD
ALA151309-100G ALA151309-1G ALA151309250MG ALA151309-25G ALA151309-5G
7-Aminoenanthic acid ≥98% (by titration)
7-Aminoenanthic acid
Formula: C₇H₁₅NO₂
MW: 145.2 g/mol
Melting Pt: 192-195°C
Storage Temperature: Ambient
MDL Number: MFCD00008242
CAS Number: 929-17-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e42bd14abb9b654d52dd787e9d7942964649b7c&vid=8e42bd14abb9b654d52dd787e9d7942964649b7c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR