4'-(4-Bromophenyl)acetophenone ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151307-100G A151307-1G A151307-25G A151307-5G
ALA151307-100GEA 169.01 USD
ALA151307-100G ALA151307-1G ALA151307-25G ALA151307-5G
4'-(4-Bromophenyl)acetophenone ≥98% (by GC)
4'-(4-Bromophenyl)acetophenone
Formula: C₁₄H₁₁BrO
Melting Pt: 127 °C
MDL Number: MFCD00143242
CAS Number: 5731-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR