1-Acetylpyrrolidine ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 1-(Pyrrolidin-1-yl)ethanone

A151303-1G A151303-25G A151303-5G
ALA151303-1GEA 44.56 USD
ALA151303-1G ALA151303-25G ALA151303-5G
1-Acetylpyrrolidine ≥97% (by GC)
1-Acetylpyrrolidine
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 225°C
Density: 1.02 g/cm³
Flash Pt: 91 °C
MDL Number: MFCD00014096
CAS Number: 4030-18-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=affb0aea16dd87d4475ec861a5a21a162676ca10&vid=affb0aea16dd87d4475ec861a5a21a162676ca10&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR