AABD-SH (=4-Acetamido-7-mercapto-2,1,3-benzoxadiazole) ≥95% (by HPLC) for HPLC labeling

Supplier: Aladdin Scientific

A151302-100MG A151302-10MG
ALA151302100MGEA 233.61 USD
ALA151302100MG ALA151302-10MG
AABD-SH (=4-Acetamido-7-mercapto-2,1,3-benzoxadiazole) ≥95% (by HPLC) for HPLC labeling
AABD-SH (=4-Acetamido-7-mercapto-2,1,3-benzoxadiazole)
Formula: C₈H₇N₃O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD04038408
CAS Number: 254973-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=193039045bcfc11c3dd4c1743c3f7add8ed479ca&vid=193039045bcfc11c3dd4c1743c3f7add8ed479ca&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR