6-Acetoxy-2-naphthoic acid ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
6-Acetoxy-2-naphthoic Acid
A151301-1G
A151301-200MG
A151301-25G
A151301-5G
ALA151301-1GEA
56.91
USD
ALA151301-1G
ALA151301200MG
ALA151301-25G
ALA151301-5G
6-Acetoxy-2-naphthoic acid ≥98%
6-Acetoxy-2-naphthoic acid
Formula:
C₁₃H₁₀O₄ MW: 230.22 g/mol Melting Pt: 228 °C |
MDL Number:
MFCD00059556 CAS Number: 17295-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...