Allyltris(trimethylsilyloxy)silane ≥96% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Allyl-1,1,1,5,5,5-hexamethyl-3-((trimethylsilyl)oxy)trisiloxane

A151298-250MG A151298-5G A151298-1G
ALA151298250MGEA 36.96 USD
ALA151298250MG ALA151298-5G ALA151298-1G
Allyltris(trimethylsilyloxy)silane ≥96% (by GC)
Allyltris(trimethylsilyloxy)silane
Formula: C₁₂H₃₂O₃Si₄
MW: 336.73 g/mol
Boiling Pt: 111 °C
Flash Pt: 67°C
Storage Temperature: Ambient
MDL Number: MFCD00054804
CAS Number: 7087-21-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b7dab8dec87b89196151a3675fae41e79f8ed2e&vid=7b7dab8dec87b89196151a3675fae41e79f8ed2e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR