Acetone thiosemicarbazone ≥97% (by titration)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: Acetone Thiosemicarbazone

A151291-100G A151291-25G
ALA151291-100GEA 107.26 USD
ALA151291-100G ALA151291-25G
Acetone thiosemicarbazone ≥97% (by titration)
Acetone thiosemicarbazone
Formula: C₄H₉N₃S
MW: 131.20 g/mol
Melting Pt: 175 °C
MDL Number: MFCD00022153
CAS Number: 1752-30-3
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fef53c59485c9d5e66f6cb141bc16d91ff1124f&vid=6fef53c59485c9d5e66f6cb141bc16d91ff1124f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR