4'-Amino-3'-(trifluoromethyl)acetanilide ≥98%

Supplier: ALADDIN SCIENTIFIC

A151289-1G A151289-25G A151289-5G
ALA151289-1GEA 9.41 USD
ALA151289-1G ALA151289-25G ALA151289-5G
4'-Amino-3'-(trifluoromethyl)acetanilide ≥98%
4'-Amino-3'-(trifluoromethyl)acetanilide
Formula: C₉H₉F₃N₂O
Melting Pt: 119 °C
Storage Temperature: Ambient
MDL Number: MFCD00270762
CAS Number: 1579-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d451efec054b988484b77fbcb003b4b1244c2722&vid=d451efec054b988484b77fbcb003b4b1244c2722&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR