4-Aminononafluorobiphenyl ≥96% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151288-5G A151288-1G A151288-200MG
ALA151288-5GEA 334.31 USD
ALA151288-5G ALA151288-1G ALA151288200MG
4-Aminononafluorobiphenyl ≥96% (by GC)
4-Aminononafluorobiphenyl
Formula: C₁₂H₂F₉N
Melting Pt: 144.0 - 148.0 °C
MDL Number: MFCD00671483
CAS Number: 969-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91714d05e702b4b2a88e2b7026c82d5af7c2f70f&vid=91714d05e702b4b2a88e2b7026c82d5af7c2f70f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR