ɑ-Cyano-3-hydroxycinnamic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Cyano-3-(3-hydroxyphenyl)acrylic acid, α-Cyano-3-hydroxycinnamic acid, 2-Cyano-3-(3-hydroxyphenyl)acrylic Acid

A151282-1G A151282-5G
ALA151282-1GEA 45.51 USD
ALA151282-1G ALA151282-5G
ɑ-Cyano-3-hydroxycinnamic acid ≥98%
alpha-Cyano-3-hydroxycinnamic acid
Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
Melting Pt: 222 °C
MDL Number: MFCD00004203
CAS Number: 54673-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c702e91ad3163ff66ac6ad17d3638062a9dc4e8&vid=6c702e91ad3163ff66ac6ad17d3638062a9dc4e8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR