3-Amino-3-(4-methylphenyl)propionic acid ≥98% (by HPLC, titration analysis)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 3-Amino-3-p-tolyl-propionic acid, DL-3-Amino-3-p-tolyl-propionic acid, 3-Amino-3-(4-methylphenyl)propionic acid, 3-Amino-3-(p-tolyl)propionic acid

A151280-1G A151280-250MG A151280-5G
ALA151280-1GEA 18.91 USD
ALA151280-1G ALA151280250MG ALA151280-5G
3-Amino-3-(4-methylphenyl)propionic acid ≥98% (by HPLC, titration analysis)
3-Amino-3-(4-methylphenyl)propionic acid
Formula: C₁₀H₁₃NO₂
MW: 179.2157 g/mol
Melting Pt: 219°C
MDL Number: MFCD01863249
CAS Number: 68208-18-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67c3ed82671ed272bf8c11b59dabd3147dd114a0&vid=67c3ed82671ed272bf8c11b59dabd3147dd114a0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR