(4-Acetylphenoxy)acetic acid ≥98% (by GC, titration analysis)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A151279-1G
A151279-25G
A151279-5G
ALA151279-1GEA
9.41
USD
ALA151279-1G
ALA151279-25G
ALA151279-5G
(4-Acetylphenoxy)acetic acid ≥98% (by GC, titration analysis)
(4-Acetylphenoxy)acetic acid
4-Acetylphenoxyacetic acid is a phenoxyacetic acid derivative. It has been synthesized by employing para-acetyl phenol as starting reagent.
Formula:
(C₆H₄(C(O)CH₃))OCH₂COOH Melting Pt: 172-177°C |
MDL Number:
MFCD00014363 CAS Number: 1878-81-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...