(3-Acetylphenoxy)acetic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-(3-Acetylphenoxy)acetic acid

A151278-1G A151278-25G A151278-5G
ALA151278-1GEA 24.61 USD
ALA151278-1G ALA151278-25G ALA151278-5G
(3-Acetylphenoxy)acetic acid ≥98%
(3-Acetylphenoxy)acetic acid
Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Melting Pt: 119 °C
MDL Number: MFCD02180947
CAS Number: 1878-80-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eaac860b99be6f4e4f7468bc6ec3a510d3696ba6&vid=eaac860b99be6f4e4f7468bc6ec3a510d3696ba6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR