Allyltrimethoxysilane ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Trimethoxyallylsilane

A151275-1ML A151275-25ML A151275-5ML
ALA151275-1MLEA 9.41 USD
ALA151275-1ML ALA151275-25ML ALA151275-5ML
Allyltrimethoxysilane ≥97% (by GC)
Allyltrimethoxysilane

Allyltrimethoxysilane is an allylating reagent that can be used for the allylation of carbonyl compounds such as aldehydes, ketones, and imines. Homoallylic alcohols and amines are obtained via the C-C bond forming reaction.It can also be used to synthesize homoallylic α-branched amines from aromatic and aliphatic aldehyde hydrazones, and ketone hydrazones.

Formula: H₂CCHCH₂Si(OCH₃)₃
MW: 162.26 g/mol
Boiling Pt: 136°C
Density: 0.964 g/cm³
Flash Pt: 31°C
Storage Temperature: Ambient
MDL Number: MFCD00053865
CAS Number: 2551-83-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0edeeee713b29ded746ab0fe54b0e90521915c43&vid=0edeeee713b29ded746ab0fe54b0e90521915c43&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR