(±)-3-Aminoquinuclidine dihydrochloride ≥98% (by titration)
Supplier: Aladdin Scientific
|
Warning
|
A151272-1G
A151272-250MG
ALA151272-1GEA
14.16
USD
ALA151272-1G
ALA151272250MG
(±)-3-Aminoquinuclidine dihydrochloride ≥98% (by titration)
(±)-3-Aminoquinuclidine dihydrochloride
Formula:
C₇H₁₆Cl₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD00137395 CAS Number: 6530-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...