4-Amino-6-chloro-2-(methylthio)pyrimidine ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A151269-1G
A151269-25G
A151269-5G
ALA151269-1GEA
15.11
USD
ALA151269-1G
ALA151269-25G
ALA151269-5G
4-Amino-6-chloro-2-(methylthio)pyrimidine ≥98% (by GC)
4-Amino-6-chloro-2-(methylthio)pyrimidine
Formula:
C₅H₆ClN₃S MW: 175.6400 g/mol Melting Pt: 133 °C |
MDL Number:
MFCD00006088 CAS Number: 1005-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...